Assay Details
| Gene Symbol: Igsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013120.1 | NP_001013138.1 | 5 - 6 | 898 | 144 | |
| GenBank mRNA | BC085929.1 | - | 5 - 6 | 898 | 7110445 | 144 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P648707 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P218305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74754 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P218305 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P648707 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P218305 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P648707 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P648707 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P218305 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P218305 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68403 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12713 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87565 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6944 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76255 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101027 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7540 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69113 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78841 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29399 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48950 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10754152 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754150 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754148 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754153 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754146 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754147 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754149 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754151 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10754145 | - | - |
| Rat Genome 230 2.0 Array | 1376733_at | - | - |
| Rat Genome 230 2.0 Array | 1376733_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376733_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376733_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100009908.rn.1 | TC1100001644.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376733_at | - | - |
| Rat Expression Set 230 | 1376733_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10754145 | - | - |
| Rat Genome U34 Set | rc_AI072647_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21798000 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650776 | - | - |

Igsf11
Genomic Map