Assay Details
| Gene Symbol: Mob3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108970.1 | NP_001102440.1 | 3 - 4 | 1036 | 103 | |
| XM_006238001.2 | XP_006238063.1 | 3 - 4 | 1049 | 103 | ||
| XM_008763636.1 | XP_008761858.1 | 3 - 4 | 1725 | 103 | ||
| GenBank mRNA | FQ220583.1 | - | 2 - 3 | 527 | 103 | |
| FQ222144.1 | - | 3 - 4 | 994 | 103 | ||
| Gene Symbol: LOC102550026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763645.1 | XP_008761867.1 | 2 - 3 | 1688 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P034849 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P034844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40162 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034849 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034844 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034849 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034844 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P034849 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P034844 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P265209 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86968 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92644 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47301 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10875955 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875956 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10875954 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500018537.rn.1 | TC0500002504.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500018541.rn.1 | TC0500002504.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500018542.rn.1 | TC0500002504.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10875954 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21116957 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363950 | - | - |

Mob3b
Genomic Map