Assay Details
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108662.1 | NP_001102132.1 | 16 - 17 | 1784 | 66 | |
XM_006238082.2 | XP_006238144.1 | 16 - 17 | 1834 | 66 | ||
XM_008763677.1 | XP_008761899.1 | 17 - 18 | 1926 | 66 | ||
XM_008763678.1 | XP_008761900.1 | 15 - 16 | 1735 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
peptidyl-tyrosine phosphorylation
positive regulation of apoptotic process
protein autophosphorylation
neural precursor cell proliferation
protein kinase activity
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
calcium ion binding
ATP binding
apoptotic process
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
peptidyl-tyrosine phosphorylation
positive regulation of apoptotic process
protein autophosphorylation
neural precursor cell proliferation
protein kinase activity
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
calcium ion binding
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P165168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79075 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165168 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P165168 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P165168 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P995681 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22233 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103284 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10868653 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868652 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868646 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868650 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868651 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868659 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868654 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868648 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868647 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868655 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868649 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868660 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868644 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868645 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868656 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868657 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868661 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10868643 | - | - |
Rat Genome 230 2.0 Array | 1389756_at | - | - |
Rat Genome 230 2.0 Array | 1389756_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389756_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389756_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500003581.rn.1 | TC0500000550.rn.1 | RTA TC |
Rat Expression Set 230 | 1389756_PM_at | - | - |
Rat Expression Set 230 | 1389756_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868643 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21023246 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364553 | - | - |
