Assay Details
Gene Symbol: Trim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107934.1 | NP_001101404.1 | 2 - 3 | 473 | 60 | |
XM_006238060.2 | XP_006238122.1 | 3 - 4 | 612 | 60 | ||
XM_006238061.2 | XP_006238123.1 | 3 - 4 | 517 | 60 | ||
XM_006238062.2 | XP_006238124.1 | 3 - 4 | 625 | 60 | ||
GenBank mRNA | BC166777.1 | - | 3 - 4 | 552 | 7934596 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
