Assay Details
Gene Symbol: Trim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107934.1 | NP_001101404.1 | 3 - 4 | 635 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P712688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95118 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P712688 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P712688 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P712688 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P302458 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P171506 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59217 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60938 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32928 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17681 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10876597 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876599 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876596 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876598 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876595 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876600 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10876594 | - | - |
Rat Genome 230 2.0 Array | 1386754_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381858_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381858_at | - | - |
Rat Genome 230 2.0 Array | 1386754_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500020006.rn.1 | TC0500002688.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500020010.rn.1 | TC0500002688.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500020009.rn.1 | TC0500002688.rn.1 | RTA TC |
Rat Expression Set 230 | 1386754_at | - | - |
Rat Expression Set 230 | 1386754_PM_at | - | - |
Rat Expression Set 230 | 1381858_PM_at | - | - |
Rat Expression Set 230 | 1381858_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876594 | - | - |
Rat Genome U34 Set | rc_AA955249_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22309876 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370734 | - | - |
