Assay Details
Gene Symbol: Rars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106643.2 | NP_001100113.1 | 3 - 4 | 233 | 61 | |
XM_006237986.2 | XP_006238048.1 | 3 - 4 | 241 | 61 | ||
GenBank mRNA | BC158744.1 | - | 3 - 4 | 233 | 9100690 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
