Assay Details
| Gene Symbol: Spag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173555.1 | NP_001167026.1 | 3 - 4 | 1013 | 97 | |
| GenBank mRNA | BC079031.1 | - | 3 - 4 | 1013 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag8
Genomic Map