Assay Details
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107919.1 | NP_001101389.1 | 8 - 9 | 1965 | 91 | |
XM_006237942.1 | XP_006238004.1 | 8 - 9 | 1972 | 91 | ||
XM_006237943.2 | XP_006238005.1 | 5 - 6 | 1655 | 91 | ||
GenBank mRNA | BC161859.1 | - | 8 - 9 | 1972 | 7096047 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P654878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130143 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P654878 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P654878 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P654878 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P286259 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15216 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10867903 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867907 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867909 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867911 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867913 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867906 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867905 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867908 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867910 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867904 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867902 | - | - |
Rat Genome 230 2.0 Array | 1391005_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392875_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391005_at | - | - |
Rat Genome 230 2.0 Array | 1392875_at | - | - |
Affymetrix HT Rat Focus Array | 1392875_at | - | - |
Affymetrix HT Rat Focus Array | 1392875_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391005_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391005_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500002020.rn.1 | TC0500000317.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002019.rn.1 | TC0500000317.rn.1 | RTA TC |
Rat Expression Set 230 | 1391005_at | - | - |
Rat Expression Set 230 | 1392875_PM_at | - | - |
Rat Expression Set 230 | 1391005_PM_at | - | - |
Rat Expression Set 230 | 1392875_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867902 | - | - |
Rat Genome U34 Set | rc_AA964134_at | - | - |
Rat Genome U34 Set | rc_AA997732_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21015003 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356157 | - | - |
