Assay Details
Gene Symbol: Gabrr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017291.3 | NP_058987.2 | 5 - 6 | 679 | 66 | |
XM_008763609.1 | XP_008761831.1 | 5 - 6 | 462 | 66 | ||
GenBank mRNA | U21070.1 | - | 5 - 6 | 636 | 66 | |
X95579.1 | - | 5 - 6 | 679 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
