Assay Details
| Gene Symbol: Nans | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106655.2 | NP_001100125.2 | 2 - 3 | 373 | 133 | |
| GenBank mRNA | BC161979.1 | - | 2 - 3 | 367 | 9029956 | 133 |
| CV116524.1 | - | 1 - 2 | 129 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nans
Genomic Map