Assay Details
| Gene Symbol: RGD1562865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135754.1 | NP_001129226.1 | 2 - 3 | 245 | 86 | |
| XM_008763617.1 | XP_008761839.1 | 1 - 2 | 338 | 86 | ||
| XM_008763618.1 | XP_008761840.1 | 2 - 3 | 327 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001135754.1, XM_008763617.1, XM_008763618.1 transcripts.
Set Membership: |
|

RGD1562865
Genomic Map