Assay Details
| Gene Symbol: RGD1562865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135754.1 | NP_001129226.1 | 3 - 4 | 496 | 110 | |
| XM_008763617.1 | XP_008761839.1 | 2 - 3 | 589 | 110 | ||
| XM_008763618.1 | XP_008761840.1 | 3 - 4 | 578 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RGD1562865
Genomic Map