Assay Details
| Gene Symbol: Perp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106265.2 | NP_001099735.1 | 1 - 2 | 316 | 103 | |
| GenBank mRNA | BC166608.1 | - | 1 - 2 | 316 | 5599021 | 103 |
More Information
Updated Mapping Information
Set Membership: |
|

Perp
Genomic Map