Assay Details
Gene Symbol: RGD1560020_predicted | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758639.1 | XP_008756861.1 | 9 - 10 | 3824 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1560020_predicted | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758639.1 | XP_008756861.1 | 9 - 10 | 3824 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |