Assay Details
| Gene Symbol: Aldh8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191088.1 | NP_001178017.1 | 3 - 4 | 484 | 86 | |
| XM_006227674.2 | XP_006227736.1 | 3 - 4 | 519 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh8a1
Genomic Map