Assay Details
| Gene Symbol: Popdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037369.1 | NP_001032446.1 | 2 - 3 | 493 | 87 | |
| GenBank mRNA | BC105761.1 | - | 2 - 3 | 493 | 5623862 | 87 |
| FQ218039.1 | - | 2 - 3 | 395 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Popdc3
Genomic Map