Assay Details
Gene Symbol: Armc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256579.2 | XP_006256641.1 | 9 - 10 | 1328 | 61 | |
XM_574794.4 | XP_574794.3 | 9 - 10 | 1241 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
