Assay Details
| Gene Symbol: Bves | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077590.1 | NP_001071058.1 | 4 - 5 | 665 | 65 | |
| GenBank mRNA | AY786529.2 | - | 4 - 5 | 665 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bves
Genomic Map