Assay Details
| Gene Symbol: Lin28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001069344.4 | XP_001069344.2 | 2 - 3 | 1623 | 117 | |
| XM_006256600.2 | XP_006256662.1 | 2 - 3 | 1057 | 117 | ||
| XM_006256601.1 | XP_006256663.1 | 3 - 4 | 488 | 117 | ||
| XM_008773029.1 | XP_008771251.1 | 3 - 4 | 831 | 117 | ||
More Information
Set Membership: |
|

Lin28b
Genomic Map