Assay Details
| Gene Symbol: Bend3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256584.2 | XP_006256646.1 | 3 - 4 | 851 | 74 | |
| XM_008773024.1 | XP_008771246.1 | 2 - 3 | 467 | 74 | ||
More Information
Set Membership: |
|

Bend3
Genomic Map