Assay Details
| Gene Symbol: Hace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108539.3 | NP_001102009.2 | 9 - 10 | 815 | 68 | |
| XM_006256595.2 | XP_006256657.1 | 9 - 10 | 1068 | 68 | ||
| XM_006256597.2 | XP_006256659.1 | 9 - 10 | 1068 | 68 | ||
| XM_006256598.1 | XP_006256660.1 | 4 - 5 | 406 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hace1
Genomic Map