Assay Details
Gene Symbol: LOC683897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109448.1 | NP_001102918.1 | 1 - 2 | 83 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109448.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
