Assay Details
Gene Symbol: LOC683897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109448.1 | NP_001102918.1 | 3 - 4 | 640 | 117 | |
XM_006256581.2 | XP_006256643.1 | 4 - 5 | 687 | 117 | ||
GenBank mRNA | BC105890.1 | - | 2 - 3 | 569 | 7306492 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
