Assay Details
| Gene Symbol: Imp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009700.1 | NP_001009700.1 | 6 - 7 | 923 | 70 | |
| GenBank mRNA | AY383672.1 | - | 591 | 70 | ||
| BC087066.1 | - | 6 - 7 | 923 | 7132412 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Imp4
Genomic Map