Assay Details
| Gene Symbol: Ptp4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031579.2 | NP_113767.1 | 1 - 2 | 186 | 101 | |
| GenBank mRNA | BC097307.1 | - | 1 - 2 | 186 | 7446277 | 101 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC097307.1,NM_031579.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ptp4a1
Genomic Map