Assay Details
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008527.1 | NP_001008527.1 | 21 - 22 | 2405 | 86 | |
XM_006245004.2 | XP_006245066.1 | 21 - 22 | 2433 | 86 | ||
XM_008767121.1 | XP_008765343.1 | 21 - 22 | 2434 | 86 | ||
XM_008767122.1 | XP_008765344.1 | 17 - 18 | 3102 | 86 | ||
XM_008767123.1 | XP_008765345.1 | 15 - 16 | 1924 | 86 | ||
XM_008767124.1 | XP_008765346.1 | 16 - 17 | 2100 | 86 | ||
GenBank mRNA | AY665586.1 | - | 21 - 22 | 2405 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P053666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158690 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053666 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053666 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P053666 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P22202 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93601 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23797 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62329 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32330 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24975 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30144 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10923464 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923434 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923441 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923453 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923443 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923437 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923439 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923454 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923452 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923445 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923436 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923442 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923448 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923444 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923466 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923433 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923463 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923459 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923469 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923440 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923455 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923457 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923435 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923449 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923446 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923465 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923438 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923467 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923468 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923447 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923451 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10923432 | - | - |
Affymetrix Rat Clariom S Assay | PSR0900005572.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005575.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005576.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005579.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005581.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005586.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005594.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005571.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005582.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005584.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005587.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005588.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005585.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005566.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005570.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005592.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005595.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005596.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005568.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005580.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005589.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005564.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005583.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005593.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900005578.rn.1 | TC0900000699.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10923432 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21666839 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363705 | - | - |
