Assay Details
| Gene Symbol: Hspd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022229.2 | NP_071565.2 | 2 - 3 | 230 | 100 | |
| XM_006244932.2 | XP_006244994.1 | 2 - 3 | 266 | 100 | ||
| GenBank mRNA | BC086507.1 | - | 2 - 3 | 230 | 7103955 | 100 |
| X53585.1 | - | 1 - 2 | 94 | 100 | ||
| X54793.1 | - | 1 - 2 | 178 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hspd1
Genomic Map