Assay Details
| Gene Symbol: LOC102549404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771480.1 | XP_008769702.1 | 3 - 4 | 424 | 113 | |
| Gene Symbol: Spock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271297.1 | NP_001258226.1 | 163 | 113 | ||
| GenBank mRNA | CB608950.1 | - | 172 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271297.1 transcripts.
Set Membership: |
|

LOC102549404
Genomic Map