Assay Details
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107336.1 | NP_001100806.1 | 2 - 3 | 399 | 135 | |
XM_006253619.2 | XP_006253681.1 | 2 - 3 | 411 | 135 | ||
GenBank mRNA | BC166772.1 | - | 2 - 3 | 391 | 7934143 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
