Assay Details
| Gene Symbol: Abcb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007796.1 | NP_001007797.1 | 9 - 10 | 1218 | 66 | |
| XM_006235946.2 | XP_006236008.1 | 8 - 9 | 1014 | 66 | ||
| GenBank mRNA | BC085781.1 | - | 9 - 10 | 1261 | 7107433 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb8
Genomic Map