Assay Details
Gene Symbol: Crispld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134963.1 | NP_001128435.1 | 4 - 5 | 1285 | 88 | |
XM_008763477.1 | XP_008761699.1 | 4 - 5 | 959 | 88 | ||
XM_008763478.1 | XP_008761700.1 | 3 - 4 | 638 | 88 | ||
GenBank mRNA | BC169118.1 | - | 4 - 5 | 924 | 9029822 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008763478.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
