Assay Details
| Gene Symbol: Chd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749899.3 | XP_003749947.1 | 19 - 20 | 4939 | 68 | |
| XM_006237848.2 | XP_006237910.1 | 19 - 20 | 4939 | 68 | ||
More Information
Set Membership: |
|

Chd7
Genomic Map