Assay Details
| Gene Symbol: Ldb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106009.1 | NP_001099479.1 | 3 - 4 | 630 | 86 | |
| XM_008770199.1 | XP_008768421.1 | 3 - 4 | 631 | 86 | ||
| XM_008770200.1 | XP_008768422.1 | 3 - 4 | 631 | 86 | ||
| XM_008770201.1 | XP_008768423.1 | 3 - 4 | 631 | 86 | ||
| XM_008770202.1 | XP_008768424.1 | 3 - 4 | 631 | 86 | ||
| XM_008770203.1 | XP_008768425.1 | 3 - 4 | 631 | 86 | ||
| GenBank mRNA | BC168881.1 | - | 3 - 4 | 515 | 8363537 | 86 |
More Information
Updated Mapping Information
Set Membership: |
|

Ldb2
Genomic Map