Assay Details
Gene Symbol: Fbxl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107222.1 | NP_001100692.1 | 9 - 10 | 1871 | 80 | |
XM_006251076.2 | XP_006251138.1 | 10 - 11 | 2060 | 80 | ||
GenBank mRNA | FQ212545.1 | - | 2 - 3 | 532 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P458395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44059 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P458395 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P458395 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P458395 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P458395 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17699 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10773149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773158 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773159 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773157 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773155 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10773148 | - | - |
Rat Genome 230 2.0 Array | 1372776_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372776_at | - | - |
Affymetrix HT Rat Focus Array | 1372776_at | - | - |
Affymetrix HT Rat Focus Array | 1372776_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400004975.rn.1 | TC1400000722.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400004974.rn.1 | TC1400000722.rn.1 | RTA TC |
Rat Expression Set 230 | 1372776_at | - | - |
Rat Expression Set 230 | 1372776_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10773148 | - | - |
Rat Genome U34 Set | rc_AA924421_at | - | - |
Rat Genome U34 Set | rc_AI013860_at | - | - |
Rat Genome U34 Set | rc_AI136252_i_at | - | - |
Rat Genome U34 Set | rc_AI136252_f_at | - | - |
Rat Genome U34 Set | rc_AI136252_r_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22358107 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368423 | - | - |
