Assay Details
Gene Symbol: LOC680039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109388.1 | NP_001102858.1 | 5 - 6 | 879 | 64 | |
XM_006251140.2 | XP_006251202.1 | 5 - 6 | 921 | 64 | ||
XM_006251141.2 | XP_006251203.1 | 5 - 6 | 921 | 64 | ||
XM_008770360.1 | XP_008768582.1 | 3 - 4 | 642 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
