Assay Details
| Gene Symbol: Evc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170439.1 | NP_001163910.1 | 17 - 18 | 2642 | 59 | |
| XM_006251090.2 | XP_006251152.1 | 17 - 18 | 2676 | 59 | ||
| XM_006251091.1 | XP_006251153.1 | 14 - 15 | 2139 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Evc
Genomic Map