Assay Details
| Gene Symbol: Cabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007730.1 | NP_001007731.1 | 2 - 3 | 697 | 74 | |
| GenBank mRNA | AY841152.1 | - | 2 - 3 | 310 | 74 | |
| BC081959.1 | - | 2 - 3 | 697 | 7124492 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cabp7
Genomic Map