Assay Details
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770223.1 | XP_008768445.1 | 5 - 6 | 938 | 72 | |
GenBank mRNA | FQ222654.1 | - | 5 - 6 | 868 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
