Assay Details
| Gene Symbol: Dhx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191597.1 | NP_001178526.1 | 7 - 8 | 1504 | 82 | |
| XM_006251031.2 | XP_006251093.1 | 7 - 8 | 1508 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhx15
Genomic Map