Assay Details
| Gene Symbol: Nacad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC091311.1 | - | 5 - 6 | 1628 | 70 | |
| BI289520.1 | - | 2 - 3 | 70 | |||
| FQ222077.1 | - | 1 - 2 | 111 | 70 | ||
More Information
Set Membership: |
|

Nacad
Genomic Map