Assay Details
Gene Symbol: Mepe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250635.2 | XP_006250697.1 | 1 - 2 | 1748 | 108 | |
GenBank mRNA | AF530559.1 | - | 1 - 2 | 90 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF530559.1, XM_006250635.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
