Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770020.1 | XP_008768242.1 | 1 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008770020.1 transcripts.
Set Membership: |
|

Agpat9
Genomic Map