Assay Details
| Gene Symbol: Helq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014134.2 | NP_001014156.2 | 7 - 8 | 1727 | 91 | |
| XM_006250666.2 | XP_006250728.1 | 7 - 8 | 1724 | 91 | ||
| XM_008770040.1 | XP_008768262.1 | 5 - 6 | 553 | 91 | ||
| XM_008770041.1 | XP_008768263.1 | 2 - 3 | 177 | 91 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_008770041.1 transcripts.
Set Membership: |
|

Helq
Genomic Map