Assay Details
| Gene Symbol: Cnot6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108355.1 | NP_001101825.1 | 3 - 4 | 450 | 82 | |
| XM_006250708.2 | XP_006250770.1 | 3 - 4 | 482 | 82 | ||
| XM_006250709.2 | XP_006250771.1 | 3 - 4 | 369 | 82 | ||
| XM_008770042.1 | XP_008768264.1 | 2 - 3 | 3249 | 82 | ||
| XM_008770043.1 | XP_008768265.1 | 2 - 3 | 261 | 82 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_008770043.1 transcripts.
Set Membership: |
|

Cnot6l
Genomic Map