Assay Details
| Gene Symbol: Cd47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248262.2 | XP_006248324.1 | 2 - 3 | 1089 | 119 | |
| XM_006248263.2 | XP_006248325.1 | 2 - 3 | 1089 | 119 | ||
| XM_006248264.2 | XP_006248326.1 | 2 - 3 | 1089 | 119 | ||
| XM_006248265.2 | XP_006248327.1 | 2 - 3 | 1089 | 119 | ||
| XM_006248268.2 | XP_006248330.1 | 2 - 3 | 1287 | 119 | ||
More Information
Set Membership: |
|

Cd47
Genomic Map