Assay Details
| Gene Symbol: RGD1311458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009678.1 | NP_001009678.1 | 2 - 3 | 300 | 111 | |
| GenBank mRNA | BC088456.1 | - | 2 - 3 | 300 | 7127273 | 111 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311458
Genomic Map