Assay Details
Gene Symbol: Pcnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047846.1 | NP_001041311.1 | 4 - 5 | 429 | 93 | |
XM_006248241.2 | XP_006248303.1 | 3 - 4 | 402 | 93 | ||
XM_006248242.1 | XP_006248304.1 | 3 - 4 | 400 | 93 | ||
XM_008768630.1 | XP_008766852.1 | 3 - 4 | 461 | 93 | ||
XM_008768631.1 | XP_008766853.1 | 3 - 4 | 458 | 93 | ||
GenBank mRNA | AY325209.1 | - | 4 - 5 | 429 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
