Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105883.1 | NP_001099353.1 | 1 - 2 | 174 | 149 | |
| XM_006248335.2 | XP_006248397.1 | 1 - 2 | 212 | 149 | ||
| XM_006248337.1 | XP_006248399.1 | 1 - 2 | 212 | 149 | ||
| XM_006248338.2 | XP_006248400.1 | 1 - 2 | 212 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pvrl3
Genomic Map