Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248335.2 | XP_006248397.1 | 5 - 6 | 1117 | 88 | |
| XM_006248337.1 | XP_006248399.1 | 5 - 6 | 1117 | 88 | ||
| XM_008768723.1 | XP_008766945.1 | 5 - 6 | 1060 | 88 | ||
| XM_008768724.1 | XP_008766946.1 | 5 - 6 | 1774 | 88 | ||
More Information
Set Membership: |
|

Pvrl3
Genomic Map