Assay Details
| Gene Symbol: Nit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034126.1 | NP_001029298.1 | 2 - 3 | 173 | 94 | |
| GenBank mRNA | BC082034.1 | - | 2 - 3 | 173 | 94 | |
| BC100637.1 | - | 2 - 3 | 173 | 5621093 | 94 | |
| FQ219183.1 | - | 2 - 3 | 179 | 94 | ||
| FQ229934.1 | - | 2 - 3 | 185 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nit2
Genomic Map